3-(Hydroxymethyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 3-(Hydroxymethyl)benzeneboronic acid, (3-Hydroxymethyl)phenylboronic acid

H1244-5G H1244-1G
TCH1244-5GEA 72.68 CAD
TCH1244-5G TCH1244-1G
3-(Hydroxymethyl)phenylboronic acid (contains varying amounts of Anhydride)
3-(Hydroxymethyl)phenylboronic acid

CAS Number: 87199-15-3
MDL Number: MFCD01317846
Molecular Formula: C7H9BO3
Molecular Weight: 151.96
Form: Crystal
Color: White
Melting point (°C): 99


Other Product Names / Synonyms: 3-(Hydroxymethyl)benzeneboronic Acid

Formula: C₇H₉BO₃
MW: 151.96 g/mol
Melting Pt: 99 °C
MDL Number: MFCD01317846
CAS Number: 87199-15-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ae01dcd2b751ea73bae127a52e26ca9a468c647&vid=2ae01dcd2b751ea73bae127a52e26ca9a468c647&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR