(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
H1241-1G
H1241-5G
TCH1241-1GEA
91.64
CAD
TCH1241-1G
TCH1241-5G
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone ≥98.0% (by GC, total nitrogen)
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone
CAS Number: 66673-40-3
MDL Number: MFCD00077791
Molecular Formula: C5H9NO2
Molecular Weight: 115.13
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 84
Specific rotation [a]20/D: -31 deg (C=5, EtOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: D-Pyroglutaminol
Formula:
C₅H₉NO₂ Melting Pt: 84 °C |
MDL Number:
MFCD00077791 CAS Number: 66673-40-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...