1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol ≥97.0%
Supplier: TCI America
H1233-1G
H1233-5G
TCH1233-1GEA
39.5
CAD
TCH1233-1G
TCH1233-5G
1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol ≥97.0%
1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol
CAS Number: 754-96-1
MDL Number: MFCD00192200
Molecular Formula: C10H6F16O2
Molecular Weight: 462.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 132
Melting point (°C): 138
Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Monomers
Other Product Names / Synonyms: 1H,1H,10H,10H-Perfluoro-1,10-decanediol
Formula:
C₁₀H₆F₁₆O₂ Boiling Pt: 132 °C Melting Pt: 138 °C |
MDL Number:
MFCD00192200 CAS Number: 754-96-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...