4-Hydroxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid

H1228-1G H1228-5G
TCH1228-1GEA 70.74 CAD
TCH1228-1G TCH1228-5G
4-Hydroxyphenylboronic acid (contains varying amounts of Anhydride)
4-Hydroxyphenylboronic acid

CAS Number: 71597-85-8
MDL Number: MFCD01074628
Molecular Formula: C6H7BO3
Molecular Weight: 137.93
Form: Crystal
Color: White
Melting point (°C): 232


Other Product Names / Synonyms: 4-Hydroxybenzeneboronic Acid

Formula: C₆H₇BO₃
MW: 137.93 g/mol
Melting Pt: 232 °C
MDL Number: MFCD01074628
CAS Number: 71597-85-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17a18701441a9ab2809989f92f4e6fa4c4f94be0&vid=17a18701441a9ab2809989f92f4e6fa4c4f94be0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR