Hexadecan-16-olide ≥97.0%

Supplier: TCI America

Synonyms: Oxa-2-cycloheptadecanone, Hexadecanolactone, Oxacycloheptadecan-2-one, 16-Hexadecanolide

H1219-25G H1219-5G
TCH1219-25GEA 314.1 CAD
TCH1219-25G TCH1219-5G
Hexadecan-16-olide ≥97.0%
Hexadecan-16-olide

CAS Number: 109-29-5
MDL Number: MFCD00039668
Molecular Formula: C16H30O2
Molecular Weight: 254.41
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 188
Flash Point (°C): 100
Freezing point (°C): 33


Product Categories: Materials Chemistry, Monomers, Bioscience, Lipids
Other Product Names / Synonyms: 16-Hexadecanolactone

Formula: C₁₆H₃₀O₂
MW: 254.41 g/mol
Boiling Pt: 188 °C
Flash Pt: 100 °C
MDL Number: MFCD00039668
CAS Number: 109-29-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ea9cc81315714fde6b54f579acee6c11a1f42cc&vid=6ea9cc81315714fde6b54f579acee6c11a1f42cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR