4-(Hydroxymethyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 4-(Hydroxymethyl)benzeneboronic acid

H1204-1G H1204-5G H1204-25G
TCH1204-1GEA 77.42 CAD
TCH1204-1G TCH1204-5G TCH1204-25G
4-(Hydroxymethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Hydroxymethyl)phenylboronic acid

CAS Number: 59016-93-2
MDL Number: MFCD00792672
Molecular Formula: C7H9BO3
Molecular Weight: 151.96
Form: Crystal
Color: White


Other Product Names / Synonyms: 4-(Hydroxymethyl)benzeneboronic Acid

Formula: C₇H₉BO₃
MW: 151.96 g/mol
MDL Number: MFCD00792672
CAS Number: 59016-93-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fa2ed9d12b43db2b70a3ba35e470439916844d2&vid=3fa2ed9d12b43db2b70a3ba35e470439916844d2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR