3-Hydroxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
H1185-5G
H1185-1G
TCH1185-5GEA
96.38
CAD
TCH1185-5G
TCH1185-1G
3-Hydroxyphenylboronic acid (contains varying amounts of Anhydride)
3-Hydroxyphenylboronic acid
CAS Number: 87199-18-6
MDL Number: MFCD01074603
Molecular Formula: C6H7BO3
Molecular Weight: 137.93
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 213
Other Product Names / Synonyms: 3-Hydroxybenzeneboronic Acid
Formula:
C₆H₇BO₃ Melting Pt: 213 °C |
MDL Number:
MFCD01074603 CAS Number: 87199-18-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...