2-Hydroxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
H1184-1G
H1184-5G
TCH1184-1GEA
102.7
CAD
TCH1184-1G
TCH1184-5G
2-Hydroxyphenylboronic acid (contains varying amounts of Anhydride)
2-Hydroxyphenylboronic acid
CAS Number: 89466-08-0
MDL Number: MFCD01074581
Molecular Formula: C6H7BO3
Molecular Weight: 137.93
Form: Crystal
Color: White
Melting point (°C): 185
Other Product Names / Synonyms: 2-Hydroxybenzeneboronic Acid
Formula:
C₆H₇BO₃ Melting Pt: 185 °C |
MDL Number:
MFCD01074581 CAS Number: 89466-08-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...