3-(1-Piperidinylmethyl)phenol ≥98.0%

Supplier: TCI America

Synonyms: 3-(Piperidin-1-ylmethyl)phenol

H1183-5G H1183-25G
TCH1183-5GEA 167.48 CAD
TCH1183-5G TCH1183-25G
3-(1-Piperidinylmethyl)phenol ≥98.0%
3-(1-Piperidinylmethyl)phenol

CAS Number: 73279-04-6
MDL Number: MFCD00800250
Molecular Formula: C12H17NO
Molecular Weight: 191.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 139


Other Product Names / Synonyms: 3-(1-Piperidinomethyl)phenol

Formula: C₁₂H₁₇NO
Melting Pt: 139 °C
MDL Number: MFCD00800250
CAS Number: 73279-04-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b361d8a3b989d674b943ecb196084f9fea58acc&vid=7b361d8a3b989d674b943ecb196084f9fea58acc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR