2-Hydroxy-5-nitro-3-picoline ≥98.0%
Supplier: TCI America
Synonyms:
3-Methyl-5-nitropyridin-2-ol, 3-Methyl-5-nitropyridin-2(1H)-one, 3-Methyl-5-nitro-2-pyridone, 2-Hydroxy-3-methyl-5-nitropyridine
H1173-5G
H1173-25G
TCH1173-5GEA
77.42
CAD
TCH1173-5G
TCH1173-25G
2-Hydroxy-5-nitro-3-picoline ≥98.0%
2-Hydroxy-5-nitro-3-picoline
CAS Number: 21901-34-8
Molecular Formula: C6H6N2O3
Molecular Weight: 154.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 229
Other Product Names / Synonyms: 2-Hydroxy-5-nitro-3-picoline, 3-Methyl-5-nitro-2-pyridinol, 3-Methyl-5-nitro-2-pyridone
Formula:
C₆H₆N₂O₃ MW: 154.13 g/mol Melting Pt: 229 °C |
MDL Number:
MFCD03095073 CAS Number: 21901-34-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...