2-Hydroxy-5-methylpyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
5-Methyl-2-pyridone, 2-hydroxy-5-picoline
H1167-5G
H1167-25G
TCH1167-5GEA
36.34
CAD
TCH1167-5G
TCH1167-25G
2-Hydroxy-5-methylpyridine ≥98.0% (by GC, titration analysis)
2-Hydroxy-5-methylpyridine
CAS Number: 1003-68-5
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 184
Other Product Names / Synonyms: 6-Hydroxy-3-picoline, 5-Methyl-2-pyridinol, 5-Methyl-2(1H)-pyridinone
Formula:
C₆H₇NO MW: 109.13 g/mol Melting Pt: 184 °C |
MDL Number:
MFCD03092888 CAS Number: 1003-68-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...