2-Hydroxy-5-nitro-6-picoline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
H1160-5G
H1160-1G
TCH1160-5GEA
142.2
CAD
TCH1160-5G
TCH1160-1G
2-Hydroxy-5-nitro-6-picoline ≥98.0% (by GC, titration analysis)
2-Hydroxy-5-nitro-6-picoline
CAS Number: 28489-45-4
Molecular Formula: C6H6N2O3
Molecular Weight: 154.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 236
Other Product Names / Synonyms: 6-Hydroxy-3-nitro-2-picoline, 6-Methyl-5-nitro-2-pyridinol
Formula:
C₆H₆N₂O₃ Melting Pt: 236 °C |
MDL Number:
MFCD00092010 CAS Number: 28489-45-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...