1-Hexyl-3-methylimidazolium hexafluorophosphate ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
Synonyms:
1-Hexyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)
H1098-25G
H1098-5G
TCH1098-25GEA
289.14
CAD
TCH1098-25G
TCH1098-5G
1-Hexyl-3-methylimidazolium hexafluorophosphate ≥98.0% (by total nitrogen and titration analysis)
1-Hexyl-3-methylimidazolium hexafluorophosphate
CAS Number: 304680-35-1
Molecular Formula: C10H19F6N2P
Molecular Weight: 312.24
Purity/Analysis Method: >98.0% (N,T)
Form: Clear Liquid
Formula:
C₁₀H₁₉F₆N₂P MW: 312.24 g/mol |
MDL Number:
MFCD03093296 CAS Number: 304680-35-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...