1-Hexyl-3-methylimidazolium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1-n-Hexyl-3-methylimidazolium chloride

H1097-5G H1097-25G
TCH1097-5GEA 69.52 CAD
TCH1097-5G TCH1097-25G
1-Hexyl-3-methylimidazolium chloride ≥98.0% (by titrimetric analysis)
1-Hexyl-3-methylimidazolium chloride

CAS Number: 171058-17-6
Molecular Formula: C10H19ClN2
Molecular Weight: 202.73
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid

Formula: C₁₀H₁₉ClN₂
MW: 202.73
Melting Pt: -85
Density: 1.05
MDL Number: MFCD03093289
CAS Number: 171058-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84528dffd746794451e2c3ae70e6178b02476dac&vid=84528dffd746794451e2c3ae70e6178b02476dac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR