2-Hydroxy-5-nitro-4-picoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H1050-5G H1050-1G
TCH1050-5GEA 157.05 CAD
TCH1050-5G TCH1050-1G
2-Hydroxy-5-nitro-4-picoline ≥98.0% (by GC, titration analysis)
2-Hydroxy-5-nitro-4-picoline

CAS Number: 21901-41-7
MDL Number: MFCD00010690
Molecular Formula: C6H6N2O3
Molecular Weight: 154.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 190


Other Product Names / Synonyms: 2-Hydroxy-5-nitro-4-picoline, 4-Methyl-5-nitro-2-pyridinol, 4-Methyl-5-nitro-2(1H)-pyridinone, 4-Methyl-5-nitro-2-pyridone, 5-Nitro-4-picolin-2-ol

Formula: C₆H₆N₂O₃
Melting Pt: 190 °C
MDL Number: MFCD00010690
CAS Number: 21901-41-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7deb3e0909a15da7ccf1492e907e25b056491ae7&vid=7deb3e0909a15da7ccf1492e907e25b056491ae7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR