N-Hydroxy-1,8-naphthalimide ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

H1040-5G H1040-25G
TCH1040-5GEA 71.1 CAD
TCH1040-5G TCH1040-25G
N-Hydroxy-1,8-naphthalimide ≥97.0% (by HPLC, titration analysis)
N-Hydroxy-1,8-naphthalimide

CAS Number: 7797-81-1
MDL Number: MFCD00065062
Molecular Formula: C12H7NO3
Molecular Weight: 213.19
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: N-Hydroxy-1,8-naphthalenedicarboximide

Formula: C₁₂H₇NO₃
MDL Number: MFCD00065062
CAS Number: 7797-81-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e27adef6ec341622d2616aeb3a90f4b32b8b94d&vid=7e27adef6ec341622d2616aeb3a90f4b32b8b94d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR