5-Hydroxy-2-nitrobenzaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H1039-5G H1039-25G
TCH1039-5GEA 186.76 CAD
TCH1039-5G TCH1039-25G
5-Hydroxy-2-nitrobenzaldehyde ≥98.0% (by GC, titration analysis)
5-Hydroxy-2-nitrobenzaldehyde

CAS Number: 42454-06-8
MDL Number: MFCD00007332
Molecular Formula: C7H5NO4
Molecular Weight: 167.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 172


Other Product Names / Synonyms: 2-Nitro-5-hydroxybenzaldehyde

Formula: C₇H₅NO₄
Melting Pt: 172 °C
MDL Number: MFCD00007332
CAS Number: 42454-06-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef3601dab37aa8144f5f1104bfc5f3901d76e60f&vid=e7a68b7f3781f05b42b78e7254d95052de072fb5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR