L-Homoserine ≥98.0%
Supplier: TCI America
Synonyms:
H-HomoSer-OH, H-HoSer-OH, H-Hser-OH, (S)-(-)-Homoserine, L(-)-Homoserine
H1030-1G
H1030-5G
TCH1030-1GEA
99.68
CAD
TCH1030-1G
TCH1030-5G
L-Homoserine ≥98.0%
L-Homoserine
CAS Number: 672-15-1
MDL Number: MFCD00063090
Molecular Formula: C4H9NO3
Molecular Weight: 119.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 194
Specific rotation [a]20/D: 8 deg (C=2, H2O)
Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-2-Amino-4-hydroxybutyric Acid
Formula:
C₄H₉NO₃ MW: 119.12 g/mol Melting Pt: 194 °C |
MDL Number:
MFCD00063090 CAS Number: 672-15-1 Merck Index: 13,04760 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...