6-Hydroxyflavanone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H1027-5G H1027-1G
TCH1027-5GEA 259.12 CAD
TCH1027-5G TCH1027-1G
6-Hydroxyflavanone ≥98.0% (by GC, titration analysis)
6-Hydroxyflavanone

CAS Number: 4250-77-5
MDL Number: MFCD00017485
Molecular Formula: C15H12O3
Molecular Weight: 240.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 219

Formula: C₁₅H₁₂O₃
Melting Pt: 219 °C
MDL Number: MFCD00017485
CAS Number: 4250-77-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fbc862f199e51338cee10471d48ebb996949904&vid=6fbc862f199e51338cee10471d48ebb996949904&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR