4-Hydroxy-3,5-dimethylbenzaldehyde ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H1023-5G H1023-25G
TCH1023-5GEA 44.24 CAD
TCH1023-5G TCH1023-25G
4-Hydroxy-3,5-dimethylbenzaldehyde ≥98.0% (by titrimetric analysis)
4-Hydroxy-3,5-dimethylbenzaldehyde

CAS Number: 2233-18-3
MDL Number: MFCD00006946
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 117

Formula: C₉H₁₀O₂
Melting Pt: 117 °C
MDL Number: MFCD00006946
CAS Number: 2233-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49b1509fc13444aedc5bd3194e54987dc64ddf02&vid=12a2d2e70adef9c6ccc40349dce18c4d66b48b90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR