2-Hydroxyquinoline-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Hydroxycinchoninic Acid

H1016-25G H1016-5G
TCH1016-25GEA 203.35 CAD
TCH1016-25G TCH1016-5G
2-Hydroxyquinoline-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
2-Hydroxyquinoline-4-carboxylic acid

CAS Number: 15733-89-8
MDL Number: MFCD00023942
Molecular Formula: C10H7NO3
Molecular Weight: 189.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 1,2-Dihydro-2-oxoquinoline-4-carboxylic Acid, 2-Hydroxycinchoninic Acid

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
MDL Number: MFCD00023942
CAS Number: 15733-89-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c7af39f233fe23d5b039fd6b81ff8d12fa521e2&vid=641b016119ca4bc6af5615d86f1672d3356a59b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR