2-Hydroxy-3-nitropyridine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
3-Nitropyridin-2(1H)-one, 3-Nitro-2-pyridinol
H0999-5G
H0999-25G
TCH0999-5GEA
48.98
CAD
TCH0999-5G
TCH0999-25G
2-Hydroxy-3-nitropyridine ≥98.0% (by HPLC, titration analysis)
2-Hydroxy-3-nitropyridine
CAS Number: 6332-56-5
MDL Number: MFCD00006270
Molecular Formula: C5H4N2O3
Molecular Weight: 140.10
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 226
Other Product Names / Synonyms: 3-Nitro-2-pyridinol, 3-Nitro-2(1H)-pyridone
Formula:
C₅H₄N₂O₃ MW: 140.1 Melting Pt: 226 °C |
MDL Number:
MFCD00006270 CAS Number: 6332-56-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...