2-(Hydroxymethyl)-18-crown-6 ≥93.0%

Supplier: TCI America

H0990-1G H0990-200MG
TCH0990-1GEA 1568.94 CAD
TCH0990-1G TCH0990-200MG
2-(Hydroxymethyl)-18-crown-6 ≥93.0%
2-(Hydroxymethyl)-18-crown-6

CAS Number: 70069-04-4
MDL Number: MFCD00188046
Molecular Formula: C13H26O7
Molecular Weight: 294.34
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Flash Point (°C): 110
Specific Gravity (20/20): 1.16


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 1,4,7,10,13,16-Hexaoxacyclooctadecane-2-methanol

Formula: C₁₃H₂₆O₇
Boiling Pt: 208 °C
Density: 1.16 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00188046
CAS Number: 70069-04-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e742a4bcfdad22586015b930acbeb368ab119493&vid=e742a4bcfdad22586015b930acbeb368ab119493&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR