(S)-2-Amino-4-phenylbutanoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: H-HoPhe-OH, H-Homophe-OH, (S)-2-Amino-4-phenylbutyric Acid

H0985-100MG H0985-1G
TCH0985-100MGEA 30.02 CAD
TCH0985-100MG TCH0985-001G
(S)-2-Amino-4-phenylbutanoic acid ≥98.0% (by titrimetric analysis)
(S)-2-Amino-4-phenylbutanoic acid

CAS Number: 943-73-7
MDL Number: MFCD00002619
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: 44 deg (C=1, 3mol/L HCl)


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-2-Amino-4-phenylbutyric Acid

Formula: C10H13NO2
MW: 179.22 g/mol
MDL Number: MFCD00002619
CAS Number: 943-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8d65c1dc243b0e4452cb4e44ca06c56769c5a43&vid=f8d65c1dc243b0e4452cb4e44ca06c56769c5a43&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR