D-Homophenylalanine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
H0984-1G
H0984-100MG
TCH0984-001GEA
112.34
CAD
TCH0984-001G
TCH0984-100MG
D-Homophenylalanine ≥98.0% (by titrimetric analysis)
D-Homophenylalanine
CAS Number: 82795-51-5
MDL Number: MFCD00063091
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -45 deg (C=1, 3mol/L HCl)
Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (R)-2-Amino-4-phenylbutyric Acid
Formula:
C₁₀H₁₃NO₂ |
MDL Number:
MFCD00063091 CAS Number: 82795-51-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...