2-(Hydroxymethyl)-15-crown-5 ≥96.0%
Supplier: TCI America
Synonyms:
2-(Hydroxymethyl)-15-crown-5-ether
H0983-5G
H0983-25G
H0983-1G
TCH0983-005GEA
485.39
CAD
TCH0983-005G
TCH0983-25G
TCH0983-001G
2-(Hydroxymethyl)-15-crown-5 ≥96.0%
2-(Hydroxymethyl)-15-crown-5
CAS Number: 75507-25-4
MDL Number: MFCD00192215
Molecular Formula: C11H22O6
Molecular Weight: 250.29
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 107
Flash Point (°C): 110
Specific Gravity (20/20): 1.17
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 1,4,7,10,13-Pentaoxacyclopentadecane-2-methanol
Formula:
C₁₁H₂₂O₆ MW: 250.29 Boiling Pt: 107 °C Density: 1.17 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00192215 CAS Number: 75507-25-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...