2-(Hydroxymethyl)-12-crown-4 ≥95.0%

Supplier: TCI America

H0982-5G H0982-1G
TCH0982-005GEA 3856.78 CAD
TCH0982-005G TCH0982-001G
2-(Hydroxymethyl)-12-crown-4 ≥95.0%
2-(Hydroxymethyl)-12-crown-4

CAS Number: 75507-26-5
MDL Number: MFCD00067524
Molecular Formula: C9H18O5
Molecular Weight: 206.24
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 1.18


Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 1,4,7,10-Tetraoxacyclododecane-2-methanol

Formula: C₉H₁₈O₅
Density: 1.18 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00067524
CAS Number: 75507-26-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=297f1869cd13a8d815fd8baa8371ab655e6147ab&vid=297f1869cd13a8d815fd8baa8371ab655e6147ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR