L-Histidine methyl ester dihydrochloride ≥98.0%

Supplier: TCI America

Synonyms: L(+)-Histidine methyl ester dihydrochloride, H-His-OMe.2HCl, L-Methyl histidinate dihydrochloride, H-His-OMe·2HCl

H0977-25G H0977-5G
TCH0977-025GEA 120.08 CAD
TCH0977-025G TCH0977-005G
L-Histidine methyl ester dihydrochloride ≥98.0%
L-Histidine methyl ester dihydrochloride

CAS Number: 7389-87-9
MDL Number: MFCD00012701
Molecular Formula: C7H11N3O2
Molecular Weight: 242.10
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 10 deg (C=2, H2O)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-His-OMe.2HCl

Formula: C₇H₁₁N₃O₂
MW: 242.10 g/mol
MDL Number: MFCD00012701
CAS Number: 7389-87-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26008fb4b3d056f59fcec9a05305d0d6582f8cc0&vid=26008fb4b3d056f59fcec9a05305d0d6582f8cc0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR