2-(2-Hydroxyphenyl)benzothiazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-(1,3-Benzothiazol-2-yl)phenol
H0973-5G
H0973-25G
TCH0973-5GEA
59.49
CAD
TCH0973-5G
TCH0973-25G
2-(2-Hydroxyphenyl)benzothiazole ≥98.0% (by HPLC, titration analysis)
2-(2-Hydroxyphenyl)benzothiazole
CAS Number: 3411-95-8
MDL Number: MFCD00022869
Molecular Formula: C13H9NOS
Molecular Weight: 227.28
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 133
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 2-(2-Benzothiazolyl)phenol
Formula:
C₁₃H₉NOS MW: 227.29 Melting Pt: 133 °C |
MDL Number:
MFCD00022869 CAS Number: 3411-95-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...