4-Hydroxy-3-pyridinesulfonic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

H0963-5G H0963-25G
TCH0963-005GEA 61.62 CAD
TCH0963-005G TCH0963-025G
4-Hydroxy-3-pyridinesulfonic acid ≥98.0% (by HPLC, titration analysis)
4-Hydroxy-3-pyridinesulphonic acid

CAS Number: 51498-37-4
MDL Number: MFCD02093473
Molecular Formula: C5H5NO4S
Molecular Weight: 175.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Flash Point (°C): 100


Other Product Names / Synonyms: 4-Pyridinol-3-sulfonic Acid

Formula: C₅H₅NO₄S
Flash Pt: 100 °C
MDL Number: MFCD02093473
CAS Number: 51498-37-4
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6a2de55f0cd2bc4e9a52c68bb0766acfea0a8be&vid=a6a2de55f0cd2bc4e9a52c68bb0766acfea0a8be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR