Bis(cyclopentadienyl)hafnium dichloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Hafnocene dichloride

H0914-25G H0914-5G H0914-1G
TCH0914-25GEA 1161.3 CAD
TCH0914-25G TCH0914-5G TCH0914-1G
Bis(cyclopentadienyl)hafnium dichloride ≥98.0% (by titrimetric analysis)
Bis(cyclopentadienyl)hafnium dichloride

CAS Number: 12116-66-4
MDL Number: MFCD00001438
Molecular Formula: C10H10Cl2Hf
Molecular Weight: 379.58
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 233


Product Categories: Synthetic Organic Chemistry, Glycosylation, Materials Chemistry, Polymerization Catalysts, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: Bis(cyclopentadienyl)hafnium Dichloride

Formula: C₁₀H₁₀Cl₂Hf
MW: 379.58 g/mol
Melting Pt: 233 °C
MDL Number: MFCD00001438
CAS Number: 12116-66-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a7f5d05b9470dbb78c466238d804f50278eedcb&vid=7a7f5d05b9470dbb78c466238d804f50278eedcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR