Helicin ≥98.0% (by HPLC)

Supplier: TCI America

H0908-5G H0908-1G
TCH0908-5GEA 464.52 CAD
TCH0908-5G TCH0908-001G
Helicin ≥98.0% (by HPLC)
Helicin

CAS Number: 618-65-5
MDL Number: MFCD00006589
Molecular Formula: C13H16O7
Molecular Weight: 284.26
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -62 deg (C=0.8, H2O)


Product Categories: Bioscience, Glycosides, Glucose, Glycosides
Other Product Names / Synonyms: 2-Formylphenyl beta-D-Glucopyranoside, 2-(beta-D-Glucopyranosyloxy)benzaldehyde, Salicylaldehyde-beta-D-glucoside

Formula: C₁₃H₁₆O₇
MDL Number: MFCD00006589
CAS Number: 618-65-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dc043fcfd6b28f7236ca711e7fb163eabdc9e72&vid=6663ad53298644202082a5c5e6a320686723571a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR