3-Hydroxy-o-toluic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-Hydroxy-2-methylbenzoic acid

H0885-1G H0885-5G H0885-25G
TCH0885-001GEA 33.18 CAD
TCH0885-001G TCH0885-005G TCH0885-25G
3-Hydroxy-o-toluic acid ≥98.0% (by titrimetric analysis)
3-Hydroxy-o-toluic acid

CAS Number: 603-80-5
MDL Number: MFCD00671541
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 145


Other Product Names / Synonyms: 3-Hydroxy-o-toluic Acid

Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 145 °C
MDL Number: MFCD00671541
CAS Number: 603-80-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33447f7ebfd70954ca84ea91bb4e34158843b29b&vid=33447f7ebfd70954ca84ea91bb4e34158843b29b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR