5-Methoxysalicylaldehyde ≥96.0%

Supplier: TCI America

Synonyms: 6-Hydroxy-m-anisaldehyde, 2-Hydroxy-5-methoxybenzaldehyde

H0881-25G H0881-5G
TCH0881-25GEA 316 CAD
TCH0881-25G TCH0881-5G
5-Methoxysalicylaldehyde ≥96.0%
5-Methoxysalicylaldehyde

CAS Number: 672-13-9
MDL Number: MFCD00003332
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 248
Flash Point (°C): 110
Freezing point (°C): 14
Specific Gravity (20/20): 1.22


Other Product Names / Synonyms: 6-Hydroxy-m-anisaldehyde, 5-Methoxysalicylaldehyde

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 248 °C
Density: 1.22 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00003332
CAS Number: 672-13-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8609b4dd7c5f884f12d8393e8632844181e46ca&vid=feb74e6213abe502ecd000bc64d6cda8a861b5d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR