(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone ≥98.0%

Supplier: TCI America

H0867-1G H0867-5G
TCH0867-001GEA 48.98 CAD
TCH0867-001G TCH0867-005G
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone ≥98.0%
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone

CAS Number: 17342-08-4
MDL Number: MFCD00077792
Molecular Formula: C5H9NO2
Molecular Weight: 115.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 86
Specific rotation [a]20/D: 31 deg (C=5, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: L-Pyroglutaminol

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 86 °C
MDL Number: MFCD00077792
CAS Number: 17342-08-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8411613fa3393b65334d9a6d987eca24ef0da894&vid=8411613fa3393b65334d9a6d987eca24ef0da894&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR