cis-4-Hepten-1-ol ≥95.0%
Supplier: TCI America
H0833-25ML
H0833-5ML
TCH0833-25MLEA
475.58
CAD
TCH0833-25ML
TCH0833-5ML
cis-4-Hepten-1-ol ≥95.0%
cis-4-Hepten-1-ol
CAS Number: 6191-71-5
MDL Number: MFCD00015575
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 73
Specific Gravity (20/20): 0.85
Formula:
C₇H₁₄O Boiling Pt: 85 °C Density: 0.85 (20 °C) Flash Pt: 73 °C |
MDL Number:
MFCD00015575 CAS Number: 6191-71-5 UN: 1987 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...