meso-Hydrobenzoin ≥98.0%
Supplier: TCI America
H0816-25G
H0816-5G
TCH0816-25GEA
325.34
CAD
TCH0816-25G
TCH0816-005G
meso-Hydrobenzoin ≥98.0%
meso-Hydrobenzoin
CAS Number: 579-43-1
MDL Number: MFCD00064253
Molecular Formula: C14H14O2
Molecular Weight: 214.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 136
Other Product Names / Synonyms: meso-1,2-Diphenyl-1,2-ethanediol
Formula:
C₁₄H₁₄O₂ Melting Pt: 136 °C |
MDL Number:
MFCD00064253 CAS Number: 579-43-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...