(+/-)-Hydrobenzoin ≥98.0%
Supplier: TCI America
H0815-5G
H0815-1G
TCH0815-5GEA
186.44
CAD
TCH0815-5G
TCH0815-001G
(+/-)-Hydrobenzoin ≥98.0%
(+/-)-Hydrobenzoin
CAS Number: 655-48-1
MDL Number: MFCD00136059
Molecular Formula: C14H14O2
Molecular Weight: 214.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 123
Other Product Names / Synonyms: (+/-)-1,2-Diphenyl-1,2-ethanediol
Formula:
C₁₄H₁₄O₂ Melting Pt: 123 °C |
MDL Number:
MFCD00136059 CAS Number: 655-48-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...