4-Cyano-4'-heptylbiphenyl ≥95.0%

Supplier: TCI America

Synonyms: 4-Cyano-4'-n-heptylbiphenyl, 4'-Heptyl-[1,1'-biphenyl]-4-carbonitrile

H0812-1G H0812-5G
TCH0812-1GEA 120.08 CAD
TCH0812-1G TCH0812-5G
4-Cyano-4'-heptylbiphenyl ≥95.0%
4-Cyano-4'-heptylbiphenyl

CAS Number: 41122-71-8
MDL Number: MFCD00074915
Molecular Formula: C20H23N
Molecular Weight: 277.41
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 36
Flash Point (°C): 110


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 4'-Heptyl-4-biphenylcarbonitrile, 7CB

Formula: C₂₀H₂₃N
MW: 277.41 g/mol
Melting Pt: 36 °C
Flash Pt: 110 °C
MDL Number: MFCD00074915
CAS Number: 41122-71-8

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR