2-(Hydroxymethyl)-1,4-benzodioxane ≥98.0%

Supplier: TCI America

H0801-25G H0801-500G
TCH0801-025GEA 50.56 CAD
TCH0801-025G TCH0801-500G
2-(Hydroxymethyl)-1,4-benzodioxane ≥98.0%
2-(Hydroxymethyl)-1,4-benzodioxane

CAS Number: 3663-82-9
MDL Number: MFCD00006822
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 286
Melting point (°C): 88


Other Product Names / Synonyms: 1,4-Benzodioxane-2-methanol

Formula: C₉H₁₀O₃
Boiling Pt: 286 °C
Melting Pt: 88 °C
MDL Number: MFCD00006822
CAS Number: 3663-82-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bde9961e31549218ce204bbb41043ae784181e70&vid=bde9961e31549218ce204bbb41043ae784181e70&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR