α,α-Diphenyl-N-methyl-D-prolinol ≥98.0%

Supplier: TCI America

H0784-1G H0784-5G
TCH0784-1GEA 243.32 CAD
TCH0784-1G TCH0784-5G
α,α-Diphenyl-N-methyl-D-prolinol ≥98.0%
α,α-Diphenyl-N-methyl-D-prolinol

CAS Number: 144119-12-0
MDL Number: MFCD00145217
Molecular Formula: C18H21NO
Molecular Weight: 267.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 72
Specific rotation [a]20/D: -55 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts
Other Product Names / Synonyms: (R)-(-)-Diphenyl(1-methylpyrrolidin-2-yl)methanol

Formula: C₁₈H₂₁NO
Melting Pt: 72 °C
MDL Number: MFCD00145217
CAS Number: 144119-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99a5295f672ffa691f8dda7ff597852788be0257&vid=99a5295f672ffa691f8dda7ff597852788be0257&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR