α,α-Diphenyl-N-methyl-L-prolinol ≥98.0%
Supplier: TCI America
H0768-1G
H0768-100MG
H0768-5G
TCH0768-1GEA
300.83
CAD
TCH0768-1G
TCH0768-100MG
TCH0768-5G
α,α-Diphenyl-N-methyl-L-prolinol ≥98.0%
α,α-Diphenyl-N-methyl-L-prolinol
CAS Number: 110529-22-1
MDL Number: MFCD00145245
Molecular Formula: C18H21NO
Molecular Weight: 267.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 71
Specific rotation [a]20/D: 55 deg (C=1, CHCl3)
Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts
Other Product Names / Synonyms: (S)-(+)-Diphenyl(1-methylpyrrolidin-2-yl)methanol
Formula:
C₁₈H₂₁NO Melting Pt: 71 °C |
MDL Number:
MFCD00145245 CAS Number: 110529-22-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...