10-Hydroxybenzo[h]quinoline 98,0
Supplier: TCI America
H0738-1G
H0738-5G
TCH0738-1GEA
45.82
CAD
TCH0738-1G
TCH0738-5G
10-Hydroxybenzo[h]quinoline 98,0
10-Hydroxybenzo[h]quinoline
CAS Number: 33155-90-7
MDL Number: MFCD00142350
Molecular Formula: C13H9NO
Molecular Weight: 195.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 101
Formula:
C₁₃H₉NO MW: 195.22 g/mol Melting Pt: 101 °C |
MDL Number:
MFCD00142350 CAS Number: 33155-90-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...