Hesperetin ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

H0721-25G H0721-5G
TCH0721-25GEA 83.55 CAD
TCH0721-25G TCH0721-005G
Hesperetin ≥97.0% (by HPLC, titration analysis)
Hesperetin

CAS Number: 520-33-2
MDL Number: MFCD00075646
Molecular Formula: C16H14O6
Molecular Weight: 302.28
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 232
Specific rotation [a]20/D: -4 deg (C=1.8, EtOH)


Product Categories: Bioscience, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 3',5,7-Trihydroxy-4'-methoxyflavanone

Formula: C₁₆H₁₄O₆
Melting Pt: 232 °C
MDL Number: MFCD00075646
CAS Number: 520-33-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fffd19474f8747eab3b4b959581c959525e1bfbd&vid=fffd19474f8747eab3b4b959581c959525e1bfbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR