4-Hydroxy-6-methyl-2-pyrone ≥98.0%
Supplier: TCI America
Synonyms:
4-Hydroxy-6-methyl-2H-pyran-2-one, 3,5-Dihydroxysorbic acid d-lactone, 2-hydroxy-6-methyl-pyran-4-one
H0715-25G
H0715-500G
TCH0715-025GEA
95.68
CAD
TCH0715-025G
TCH0715-500G
4-Hydroxy-6-methyl-2-pyrone ≥98.0%
4-Hydroxy-6-methyl-2-pyrone
CAS Number: 675-10-5
MDL Number: MFCD00006641
Molecular Formula: C6H6O3
Molecular Weight: 126.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 188
Other Product Names / Synonyms: 3,5-Dihydroxysorbic Acid delta-Lactone
Formula:
C₆H₆O₃ MW: 126.11 g/mol Melting Pt: 188 °C |
MDL Number:
MFCD00006641 CAS Number: 675-10-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...