4-Bromo-4'-hydroxybiphenyl ≥99.0%

Supplier: TCI America

Synonyms: 4'-Bromo-[1,1'-biphenyl]-4-ol

H0690-5G H0690-25G
TCH0690-005GEA 99.54 CAD
TCH0690-005G TCH0690-025G
4-Bromo-4'-hydroxybiphenyl ≥99.0%
4-Bromo-4'-hydroxybiphenyl

CAS Number: 29558-77-8
MDL Number: MFCD00059076
Molecular Formula: C12H9BrO
Molecular Weight: 249.11
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 168


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals, Building Blocks for Liquid Crystals, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4'-Bromobiphenyl-4-ol, 4-(4-Bromophenyl)phenol

Formula: C₁₂H₉BrO
MW: 249.11 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00059076
CAS Number: 29558-77-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85688c2ded1f11ffe492635485db7fea09393e21&vid=85688c2ded1f11ffe492635485db7fea09393e21&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR