Methyl-4-(hydroxymethyl)benzoate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Methyl 4-(hydroxymethyl)benzoate
H0640-5G
H0640-25G
TCH0640-5GEA
92.59
CAD
TCH0640-5G
TCH0640-025G
Methyl-4-(hydroxymethyl)benzoate ≥98.0% (by titrimetric analysis)
Methyl 4-(hydroxymethyl)benzoate
CAS Number: 6908-41-4
MDL Number: MFCD00009984
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 157
Melting point (°C): 52
Flash Point (°C): 113
Other Product Names / Synonyms: 4-(Hydroxymethyl)benzoic Acid Methyl Ester
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Boiling Pt: 157 °C Melting Pt: 52 °C Flash Pt: 113 °C |
MDL Number:
MFCD00009984 CAS Number: 6908-41-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...