2-Hydroxy-n-octanoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Hydroxy-n-octanoic Acid, 2-Hydroxyoctanoic acid

H0592-25G H0592-5G
TCH0592-025GEA 519.82 CAD
TCH0592-025G TCH0592-005G
2-Hydroxy-n-octanoic acid ≥98.0% (by titrimetric analysis)
2-Hydroxy-n-octanoic acid

CAS Number: 617-73-2
MDL Number: MFCD00014410
Molecular Formula: C8H16O3
Molecular Weight: 160.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 69

Formula: C₈H₁₆O₃
MW: 160.21 g/mol
Melting Pt: 69 °C
MDL Number: MFCD00014410
CAS Number: 617-73-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b501f5c7b4b59981ccba81158be70f3fdbefa5b4&vid=b501f5c7b4b59981ccba81158be70f3fdbefa5b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR