N-Hydroxy-5-norbornene-2,3-dicarboximide ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: endo-N-Hydroxybicyclo[2.2.1]hept-5-ene-2,3-dicarboximide

H0528-250G H0528-25G
TCH0528-250GEA 305.73 CAD
TCH0528-250G TCH0528-025G
N-Hydroxy-5-norbornene-2,3-dicarboximide ≥99.0% (by titrimetric analysis)
N-Hydroxy-5-norbornene-2,3-dicarboximide

[for Peptide Synthesis]
CAS Number: 21715-90-2
MDL Number: MFCD00167583
Molecular Formula: C9H9NO3
Molecular Weight: 179.18
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 167

Formula: C₉H₉NO₃
MW: 179.18 g/mol
Melting Pt: 167 °C
MDL Number: MFCD00167583
CAS Number: 21715-90-2

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR