2-Hydroxypyrimidine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

H0480-250G H0480-25G
TCH0480-250GEA 429.76 CAD
TCH0480-250G TCH0480-025G
2-Hydroxypyrimidine hydrochloride ≥98.0% (by HPLC, titration analysis)
2-Hydroxypyrimidine hydrochloride

CAS Number: 38353-09-2
MDL Number: MFCD00012781
Molecular Formula: C4H4N2O
Molecular Weight: 132.55
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 2-Pyrimidinol Hydrochloride

Formula: C₄H₄N₂O
MDL Number: MFCD00012781
CAS Number: 38353-09-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7b223ac58cd2f0805448e62c88528193e249356&vid=e7b223ac58cd2f0805448e62c88528193e249356&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR